Learn More
TGFBI / BIGH3 Mouse anti-Human, CoraLite 594, Clone: 3E11D11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60007-100UL
Description
The TGFBI / BIGH3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TGFBI / BIGH3. This antibody recognizes human antigen. The TGFBI / BIGH3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| TGFBI / BIGH3 | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 7045 | |
| IgG2a |
| Immunofluorescence | |
| 3E11D11 | |
| Mouse | |
| Primary | |
| Human |