missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human TGF-beta 3 Recombinant Protein
Recombinant Human Transforming Growth Factor β 3 (TGFB3) is a bioactive protein intended for use in cell culture applications
£8190.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 7043 |
Molecular Weight (g/mol) | 25.4 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | TGF-β3 |
Description
Human TGF-beta 3 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
TGFB3 is involved in epithelial and endothelial cell proliferation and differentiation during development.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined in a toxicity assay using WHO Standard 98/608 for direct comparison
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
25.4 kDa | |
TGF-β3 | |
Store in refrigerator (2–8°C). | |
E. coli | |
Recombinant, Ligand | |
TGFs (Transforming Growth Factors) | |
Gibco™ |
7043 | |
Sequential Chromatography | |
Human | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Liquid | |
Immunology, Neurobiology, Stem Cell Research, Bone Research, Cardiovascular Research, Oncology | |
98 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.