missing translation for 'onlineSavingsMsg'
Learn More

TGF-beta RII Antibody (RM0132-6G11), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18428594 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428594 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428594 Supplier Novus Biologicals Supplier No. NBP122508MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

TGF-beta RII Monoclonal antibody specifically detects TGF-beta RII in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF-beta RII
Applications Western Blot
Classification Monoclonal
Clone RM0132-6G11
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbeta-RII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II
Host Species Rat
Immunogen Mouse recombinant TGF-beta RII (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7048
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.