missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta RII Antibody (RM0132-6G11), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22508CL1
This item is not returnable.
View return policy
Description
TGF-beta RII Monoclonal antibody specifically detects TGF-beta RII in Mouse samples. It is validated for Western Blot
Specifications
| TGF-beta RII | |
| Monoclonal | |
| CoraFluor 1 | |
| AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbeta-RII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
| Mouse recombinant TGF-beta RII (extracellular domain) | |
| 0.1 mL | |
| Angiogenesis, Cell Biology, Cell Cycle and Replication | |
| 7048 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
| Western Blot | |
| RM0132-6G11 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction