missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta RII Antibody (LY3022859) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28097JF585
This item is not returnable.
View return policy
Description
TGF-beta RII Monoclonal antibody specifically detects TGF-beta RII in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TGF-beta RII | |
| Monoclonal | |
| Janelia Fluor 585 | |
| AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbeta-RII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
| TBFbR2 | |
| 0.1 mL | |
| Angiogenesis, Cell Biology, Cell Cycle and Replication | |
| 7048 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| LY3022859 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction