missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta RI/ALK-5 Antibody (RM0016-3A11), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11921B
This item is not returnable.
View return policy
Description
TGF-beta RI/ALK-5 Monoclonal specifically detects TGF-beta RI/ALK-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TGF-beta RI/ALK-5 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TGFBR1 | |
| Mouse TGF-beta RI/ALK-5 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0016-3A11 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, tbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I | |
| Rat | |
| Protein G purified | |
| Angiogenesis, Phospho Specific, Protein Kinase | |
| 7046 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction