missing translation for 'onlineSavingsMsg'
Learn More

TGF-beta RI/ALK-5 Antibody (854621R) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30733695 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733695 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733695 Supplier Novus Biologicals Supplier No. FAB3025I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TGF-beta RI/ALK-5 Monoclonal antibody specifically detects TGF-beta RI/ALK-5 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF-beta RI/ALK-5
Applications ELISA
Classification Monoclonal
Clone 854621R
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, tbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TGF-β RI/ALK-5, Leu34-Glu125, Accession # P36897
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 7046
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.