missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems TGF-beta Premixed Magnetic Luminex™ Performance Assay

Product Code. p-200055751
Click to view available options
Quantity:
2 Plex
3 Plex
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18753517

Brand: R&D Systems FCSTM1703

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 3 Plex
Species Human, Mouse, Rat, Porcine
Binding Target TGF beta 1/2/3
For Use With (Application) ELISA
Gene ID (Entrez) 7040
Synonym ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.