missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF beta induced factor 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£385.00 - £587.00
Specifications
| Antigen | TGF beta induced factor 2 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18301536
|
Novus Biologicals
NBP3-17774-25UL |
25 μg |
£385.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18331767
|
Novus Biologicals
NBP3-17774-100UL |
100 μg |
£587.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGF beta induced factor 2 Polyclonal antibody specifically detects TGF beta induced factor 2 in Human samples. It is validated for ImmunofluorescenceSpecifications
| TGF beta induced factor 2 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 60436 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| homeobox protein TGIF2, TGF(beta)-induced transcription factor 2, TGF-beta-induced transcription factor 2,5'-TG-3'-interacting factor 2,5'-TG-3' interacting factor 2, TGFB-induced factor 2, TGFB-induced factor 2 (TALE family homeobox), TGFB-induced factor homeobox 2, transcription growth factor-beta-induced factor 2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RAAEMELQKQQDPSLPLLHTPIPLVSE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title