missing translation for 'onlineSavingsMsg'
Learn More

TGF beta induced factor 2 Antibody (5B11), Novus Biologicals™

Product Code. 18427469 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427469 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427469 Supplier Novus Biologicals Supplier No. H00060436M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TGF beta induced factor 2 Monoclonal antibody specifically detects TGF beta induced factor 2 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF beta induced factor 2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 5B11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_068581
Gene Alias homeobox protein TGIF2, TGF(beta)-induced transcription factor 2, TGF-beta-induced transcription factor 2,5'-TG-3'-interacting factor 2,5'-TG-3' interacting factor 2, TGFB-induced factor 2, TGFB-induced factor 2 (TALE family homeobox), TGFB-induced factor homeobox 2, transcription growth factor-beta-induced factor 2
Host Species Mouse
Immunogen TGIF2 (NP_068581, 131 a.a. ∽ 236 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SMPLHSGQGEKPAAPFPRGELESPKPLVTPGSTLTLLTRAEAGSPTGGLFNTPPPTPPEQDKEDFSSFQLLVEVALQRAAEMELQKQQDPSLPLLHTPIPLVSENP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 60436
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.