missing translation for 'onlineSavingsMsg'
Learn More

TGF-beta Antibody (TGFB/7240), Novus Biologicals™

Mouse Monoclonal Antibody

£178.00 - £351.00

Specifications

Antigen TGF-beta
Clone TGFB/7240
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18617794
View Documents
Novus Biologicals
NBP3-23695-100ug
100 μg
£351.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18630715
View Documents
Novus Biologicals
NBP3-23695-20ug
20 μg
£178.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TGF-beta Monoclonal antibody specifically detects TGF-beta in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TGF-beta
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cytokine Research, Extracellular Matrix
10 mM PBS with 0.05% BSA
7040
IgG
Protein A or G purified
TGFB/7240
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Recombinant fragment (around aa200-390) of human TGF-beta protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.