missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta Antibody (1D11.16.8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08710CL1
This item is not returnable.
View return policy
Description
TGF-beta Monoclonal antibody specifically detects TGF-beta in Bovine,Canine,Hamster,Human,Monkey,Mouse,Rat samples. It is validated for ELISA,Western Blot,Functional Assay Assay,Immunocytochemistry/Immunofluorescence
Specifications
| TGF-beta | |
| Monoclonal | |
| CoraFluor 1 | |
| CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| Bovine TGF-beta protein | |
| 0.1 mL | |
| Cytokine Research, Extracellular Matrix | |
| 7040 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA, Western Blot, Functional Assay, Immunocytochemistry/Immunofluorescence | |
| 1D11.16.8 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Canine, Hamster, Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction