Learn More
PeproTech TGF beta-3 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P317BT-50UG
Description
AA Sequence of recombinant protein: ALDTNYCFRN LEENCCVRPL YIDFRQDLGW KWVHEPKGYY ANFCSGPCPY LRSADTTHST VLGLYNTLNP EASASPCCVP QDLEPLTILY YVGRTPKVEQ LSNMVVKSCK CS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TGF-beta3. Anti-Human TGF-beta3-specific antibody was purified by affinity chromatography and then biotinylated. Western Blot: To detect Human TGF-beta3 by Western Blot analysis, this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Human TGF-beta3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P317BT-1MG will be provided as 2 x 500 μg
Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity.
Specifications
| TGF beta-3 | |
| Polyclonal | |
| Biotin | |
| TGFB3 | |
| ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7043 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P10600 | |
| TGFB3 | |
| E.coli-derived Recombinant Human TGF-beta3. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.