missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 3 Antibody (Brigham and Womens anti-LAP), PE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28029PE
This item is not returnable.
View return policy
Description
TGF-beta 3 Monoclonal antibody specifically detects TGF-beta 3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TGF-beta 3 | |
| Monoclonal | |
| PE | |
| ARVD, FLJ16571, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 | |
| LAP | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Tumor Suppressors | |
| 7043 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Brigham and Womens anti-LAP | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction