Learn More
Invitrogen™ TGF beta-2 Recombinant Superclonal™ Antibody (12HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 710276
Description
Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.
Transforming Growth Factor (TGF) betas mediate many cell to cell interactions that occur during embryonic development. Three TGF betas have been identified in mammals. TGF beta 1, TGF beta 2 and TGF beta 3 are each synthesized as precursor proteins that are very similar in that each is cleaved to yield a 112 amino acid polypeptide that remains associated with the latent portion of the molecule. The TGF beta polypeptides are multifunctional; capable of influencing cell proliferation, differentiation, and other functions in a wide range of cell types. Transformed, as well as nonneoplastic tissues, release transforming growth factors; and essentially all mammalian cells possess a specific TGF receptor. The multi modal nature of TGF beta is seen in its ability to stimulate or inhibit cellular proliferation. In general, cells of mesenchymal origin appear to be stimulated by TGF beta whereas cells of epithelial or neuroectodermal origin are inhibited by the peptide. TGF beta 1, TGF beta 2, and TGF beta 1.2 appear to be equivalent in biological activity, although there does appear to be differences in binding to certain types of receptors. TGF beta 2 is produced by many cell types and has been found in the highest concentration in porcine platelets and mammalian bone. Latent TGF beta 2 is the prominent isoform found in body fluids such as amniotic fluid, breast milk, and the aqueous and vitreous humor of the eye.
Specifications
| TGF beta-2 | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P27090, P61812 | |
| TGFB2 | |
| Recombinant protein corresponding to amino acids 331-442 of human Transforming Growth Factor Beta 2. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| 12HCLC | |
| Unconjugated | |
| TGFB2 | |
| BB105277; BSC-1 cell growth inhibitor; cetermin; glioblastoma-derived T-cell suppressor factor; G-TSF; LAP; Latency-associated peptide; LDS4; polyergin; prepro-transforming growth factor beta-2; TGF beta 2 protein; TGFB; TGFB2; TGF-B2; Tgfb-2; TGF-beta2; TGF-beta-2; TGFβ2; Transforming growth factor; transforming growth factor beta 2; transforming growth factor beta-2; Transforming growth factor beta-2 proprotein; transforming growth factor, beta 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 21808, 7042 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.