missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems BAF302
This item is not returnable.
View return policy
Description
TGF-beta 2 Polyclonal specifically detects TGF-beta 2 in Multi-Species samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| TGF-beta 2 | |
| Polyclonal | |
| Biotin | |
| BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGFB2, TGFbeta 2, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7042 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| TGFB2 | |
| Porcine platelet-derived TGF-beta 2 (R&D Systems, Catalog # 102-B2) | |
| 50 μg | |
| Primary | |
| Detects TGF-beta 2 in ELISAs and Western blots. In sandwich immunoassays, approximately 10% cross-reactivity with rhTGF-beta 1.2 and less than 0.1% cross-reactivity with rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 3 and raTGF-beta 5 is observed. | |
| Multi-species | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction