missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 2 Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AB-12-NA
This item is not returnable.
View return policy
Description
TGF-beta 2 Polyclonal specifically detects TGF-beta 2 in Multi-Species samples. It is validated for Western Blot, Simple Western, Neutralization.
Specifications
| TGF-beta 2 | |
| Polyclonal | |
| Unconjugated | |
| BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGFB2, TGFbeta 2, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 7042 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, Neutralization 0.25-1.25 ug/mL | |
| TGFB2 | |
| Porcine platelet-derived TGF-beta 2 | |
| 1 mg | |
| Primary | |
| Detects TGF-beta 2 and TGF-beta 1.2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with TGF-beta 3 and TGF-beta 1 is observed. | |
| Multi-species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction