missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB612-100
This item is not returnable.
View return policy
Description
TGF-beta 2 Monoclonal specifically detects TGF-beta 2 in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| TGF-beta 2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TGFB2 | |
| Porcine TGF-beta 1.2 | |
| 100 μg | |
| Primary | |
| Detects TGF-beta 2 from multiple species in ELISAs and Western blots. In ELISAs, no cross-reactivity or interfereence with recombinant human (rh) TGF-alpha , recombinant porcineTGF-beta 1, rhTGF-beta 1, recombinant canine TGF-beta 3, recombinant amphibian TGF-beta 5, rhTGF-beta RII, or rhTGF-beta RIII is observed. | |
| Multi-species | |
| Ascites |
| Western Blot, Immunohistochemistry, ELISA | |
| 8607 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGFB2, TGFbeta 2, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7042 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction