Learn More
Invitrogen™ TGF beta-1 Recombinant Rabbit Monoclonal Antibody (3E9Y0)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | TGF beta-1 |
|---|---|
| Clone | 3E9Y0 |
| Concentration | 0.53 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283340
|
Invitrogen™
MA558183 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30284320
![]() |
Invitrogen™
MA558184 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: LSTCKTIDME LVKRKRIEAI RGQILSKLRL ASPPSQGEVP PGPLPEAVLA LYNSTRDRVA GESADPEPEP EADYYAKEVT RVLMVDRNNA IYEKTKDISH SIYMFFNTSD IREAVPEPPL LSRAELRLQR LKSSVEQHVE LYQKYSNNSW RYLGNRLLTP TDTPEWLSFD VTGVVRQWLN QGDGIQGFRF SAHCSCDSKD NKLHVEINGI SPKRRGDLGT IHDMNRPFLL LMATPLERAQ HLHSSRHRRA LDTNY.
TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.Specifications
| TGF beta-1 | |
| 0.53 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P01137, P04202, P17246 | |
| 21803, 59086, 7040 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-390 of mouse TGFB1 (NP_035707.1). | |
| Antibody |
| 3E9Y0 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Tgfb1 | |
| CED; cytokine; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF b; TGF b 1; TGF beta; tgf beta 1; tgf beta1; TGF beta-1; TGF beta-1 protein; TGF β; TGF β 1; TGF β1; Tgfb; TGFB1; TGF-B1; Tgfb-1; TGFbeta; TGF-beta 1; TGF-beta 1 protein; TGFbeta1; TGF-beta1; TGF-beta-1; TGFβ1; Transforming growth factor; transforming growth factor b1; transforming growth factor beta; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor β1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta I; transforming growth factor-beta-1; transforming growth factor-beta-1 precursor; unnamed protein product | |
| Tgfb1 | |
| Primary | |
| Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.