missing translation for 'onlineSavingsMsg'
Learn More

TGF beta 1 Rabbit, anti-Mouse, Polyclonal, Proteintech

Product Code. 18000636 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18000636 20 μL 20µL
18016895 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18000636 Supplier Proteintech Supplier No. 261551AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The TGF beta 1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse TGF beta 1. This antibody recognizes mouse antigen. The TGF beta 1 antibody has been validated for the following applications: WB, ELISA analysis.

Specifica

Antigen TGF beta 1
Applications Western Blot, ELISA
Concentration 800 μg/mL
Gene Symbols TGF beta1, Tgfb, Tgfb 1, Tgfb1, TGF-beta 1, TGFbeta1, TGF-beta1, TGF-β, TGFβ1, TGF-β1
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 21803
Target Species Mouse
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.