missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TGF beta-1 (LAP) Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ BMS2065

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15571577

  • £451.00 / 96 tests
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human LAP

Description

Description

The Human Transforming Growth Factor Beta 1/Latency Associated Peptide (Hu TGFβ1/LAP) ELISA quantitates Hu TGFβ1/LAP in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TGFβ1/LAP. Principle of the method The Human TGFβ1/LAP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.
Specifications

Specifications

P01137
0.098 ng/mL
ELISA Kit
Human
Colorimetric Microplate Reader
CED,DPD1,LAP,TGFB,TGFbeta
7.6%
HRP
RUO
2°C to 8°C
Human
3 hr. 30 min.
0.16-10 ng/mL
Biotin
Plasma, Serum, Supernatant
ELISA
7040
6%
Pre-coated 96 well plate, Standard, Standard, Assay Buffer concentrate, Assay Buffer concentrate, SAV-HRP, SAV-HRP, Chromogen, Stop Solution, Stop Solution, Controls
96 Tests
Plasma, 10 μL; Serum, 10 μL; Supernatant, 10 μL
Latency Associated Peptide
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only