missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGF beta-1 Human ELISA Kit
For the quantitative detection of human TGF-beta1.
£451.00 - £3070.00
Specifications
Accession Number | P01137 |
---|---|
Product Type | ELISA Kit |
Sample Type | Plasma, Serum, Supernatant |
Target Species | Human |
For Use With (Application) | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15581827
|
Invitrogen™
BMS249-4TEN |
10 x 96 Tests |
£3070.00
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15561827
|
Invitrogen™
BMS249-4 |
96 Tests |
£451.00
96 tests |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
15571827
|
Invitrogen™
BMS249-4FIVE |
5 x 96 Tests |
£1548.00
Pack of 5 |
In Stock
Log in to see stock. |
|||||
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
P01137 | |
Plasma, Serum, Supernatant | |
ELISA | |
7040 | |
3.2% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 30 min. |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
4.9% | |
HRP | |
Plasma, 20 μL; Serum, 20 μL; Supernatant, 20 μL | |
Camurati-Engelmann disease, DPD1 | |
1 hr. 20 min. |