missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 1 Antibody (PD00-17), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33008
This item is not returnable.
View return policy
Description
TGF-beta 1 Monoclonal antibody specifically detects TGF-beta 1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| TGF-beta 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 | |
| CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| Synthetic peptide (KLH-coupled) corresponding to Human TGF-Beta 1 C-terminal. (Uniprot: P01137) | |
| 100 μL | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction | |
| 7040 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| PD00-17 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction