missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 1 Antibody (Genzyme patent anti-TGF-beta) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28452JF635
This item is not returnable.
View return policy
Description
TGF-beta 1 Monoclonal antibody specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TGF-beta 1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| TGFb1 | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction | |
| 7040 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genzyme patent anti-TGF-beta | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction