Learn More
Invitrogen™ TGF beta-1,2,3 Monoclonal Antibody (eBioTB2F), eBioscience™
Rat Monoclonal Antibody
£88.75 - £262.00
Specifications
| Antigen | TGF beta-1,2,3 |
|---|---|
| Clone | eBioTB2F |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15288257
|
Invitrogen™
14-9943-81 |
50 μg |
£88.75
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15298257
|
Invitrogen™
14-9943-85 |
500 μg |
£262.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The eBioTB2F antibody reacts with Transforming Growth Factor-beta (TGF-beta), a pleiotropic cytokine, exists in five isoforms, known as TGF-beta1-5. Homologies between isoforms range from 70-80% but no homology exists to TGF-alpha. TGF-beta1 is ubiquitous and the most abundant form found in lymphoid organs, while other isoforms are expressed in a more restricted distribution. The biologically active state of all isoforms are disulfide-linked homodimers. The heat- and acid- stable monomeric subunits have a length of 112 amino acids. The isoforms of TGF-beta arise by proteolytic cleavage of longer precursors; the isoforms are derived from the carboxyterminal ends of these precursors. Isoforms isolated from different species are evolutionarily closely conserved and have sequence identities on the order of 98%. Mature human, porcine, simian, chicken and bovine TGF-beta1 are identical and differ from mouse TGF-beta1 in a single amino acid. TGF-beta1 is produced in very high levels by platelets. Other cellular sources of TGF-beta1 include macrophages, lymphocytes, endothelial cells, chondrocytes, and leukemic cells. TGF-beta1 secretion can be induced by steroids, retinoids, EGF, NGF, vitamin D3, and IL-1. Activities of TGF-beta1 include inhibition of cell growth for inhibitor for normal and transformed epithelial cells, endothelial cells, fibroblasts, neurons, and lymphoid cells and other hematopoietic cell types. TGF-beta1 inhibits the proliferation of T cells and...
TGF-beta1, -2, and -3 are a closely related group of proteins (70-80% sequence homology) that are produced by many cell types and function as growth and differentiation factors. The active forms of TGF-beta1, -2, and -3 are disulfide-linked homodimers.Specifications
| TGF beta-1,2,3 | |
| 0.5 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01137, P10600, P61812 | |
| 7040, 7042, 7043 | |
| Primary | |
| Affinity chromatography |
| eBioTB2F | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human | |
| TGFB3 | |
| CED; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; tgf beta 1; tgf beta 3; tgf beta1; TGFB; Tgfb1; Tgfb-1; TGFB4; TGFbeta; TGF-beta 1; TGFbeta1; TGF-beta1; TGF-beta-1; TGF-beta4; tissue growth factor-beta4; Transforming growth factor; transforming growth factor beta 1; transforming growth factor beta 4; transforming growth factor beta 4 precursor; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta-1 precursor | |
| TGFB1, TGFB2, TGFB3 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.