missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGF beta-1,2,3 Monoclonal Antibody (eBio16TFB), Biotin, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£102.00 - £268.00
Specifications
| Antigen | TGF beta-1,2,3 |
|---|---|
| Clone | eBio16TFB |
| Concentration | 0.5 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15330120
|
Invitrogen™
13-9923-85 |
500 μg |
£268.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15320120
![]() |
Invitrogen™
13-9923-81 |
50 μg |
£102.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGF-beta1, -2, and -3 are a closely related group of proteins (70-80% sequence homology) that are produced by many cell types and function as growth and differentiation factors. The active forms of TGF-beta1, -2, and -3 are disulfide-linked homodimers.Specifications
| TGF beta-1,2,3 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CED; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; tgf beta 1; tgf beta 3; tgf beta1; TGFB; Tgfb1; Tgfb-1; TGFB4; TGFbeta; TGF-beta 1; TGFbeta1; TGF-beta1; TGF-beta-1; TGF-beta4; tissue growth factor-beta4; Transforming growth factor; transforming growth factor beta 1; transforming growth factor beta 4; transforming growth factor beta 4 precursor; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta-1 precursor | |
| TGFB1, TGFB2, TGFB3 | |
| Primary | |
| 4°C, do not freeze | |
| TGFB3 |
| eBio16TFB | |
| ELISA | |
| Biotin | |
| Mouse | |
| Human | |
| P01137, P10600, P61812 | |
| 7040, 7042, 7043 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title