missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 1, 2, 3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£203.00 - £1865.00
Specifications
| Antigen | TGF beta 1/2/3 |
|---|---|
| Clone | 1D11R |
| Dilution | Neutralization 0.25-1.25 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18720937
![]() |
R&D Systems
MAB1835R-SP |
25 μg |
£203.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18760638
![]() |
R&D Systems
MAB1835R-100 |
100 μg |
£442.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18122111
|
R&D Systems
MAB1835R-01M |
1 mg |
£1840.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109130
|
R&D Systems
MAB1835R-500 |
500 ug |
£1865.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGF beta 1/2/3 Monoclonal specifically detects TGF beta 1/2/3 in Multi-Species samples. It is validated for Neutralization.Specifications
| TGF beta 1/2/3 | |
| Neutralization 0.25-1.25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Multi-species | |
| 7040 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Detects bovine, chicken, mouse, and human TGF-beta in ELISAs and Western blots. It recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3. |
| 1D11R | |
| Neutralization | |
| Unconjugated | |
| RUO | |
| ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1 | |
| Bovine bone-derived TGF-beta 1 and TGF-beta 2 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title