missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGF alpha Recombinant Rabbit Monoclonal Antibody (JB58-36)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534680
This item is not returnable.
View return policy
Description
Positive Control: Human tonsil tissue, human liver tissue, human colon cancer tissue, human kidney tissue, A431, A549, PC-3M.
TGF-alpha is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-alpha induces anchorage-independence transformation in cultured cells. Human, murine and rat TGF-alpha are cross-species reactive. TGFalpha (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.
Specifications
| TGF alpha | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P01135 | |
| TGFA | |
| Recombinant protein within Human TGF alpha aa 1-160. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| JB58-36 | |
| Unconjugated | |
| TGFA | |
| EGF-like TGF; ETGF; I79_019067; LOC100009150; Protransforming growth factor alpha; RATTGFAA; TFGA; TGF a; TGF type 1; TGF α; TGFA; TGFAA; TGFalpha; TGF-alpha; TGFα; Transforming growth factor; Transforming growth factor alpha; transforming growth factor, alpha; transforming growth factor-alpha; wa1; wa-1; waved 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7039 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction