missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TGF-alpha Monoclonal specifically detects TGF-alpha in Human samples. It is validated for Flow Cytometry, ELISA, Dot Blot.
Specifications
Specifications
| Antigen | TGF-alpha |
| Applications | ELISA, Dot Blot |
| Classification | Monoclonal |
| Clone | tAb2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA, Dot Blot |
| Formulation | PBS |
| Gene Alias | protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha |
| Gene Symbols | TGFA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?