missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TGF alpha ELISA Kit

Product Code. 17778023
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17778023

Brand: Invitrogen™ EHTGFAX10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

TGF-alpha is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-alpha induces anchorage-independence transformation in cultured cells. Human, murine and rat TGF-alpha are cross-species reactive. TGFalpha (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01135
Assay Range 1.37 to 1000 pg/mL
Assay Sensitivity 3 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 7039
Gene Symbol TGFA
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Target TGF alpha
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.