missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-alpha Antibody (TGFA/1119), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47737F
This item is not returnable.
View return policy
Description
TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TGF-alpha | |
| Monoclonal | |
| FITC | |
| protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha | |
| Recombinant human full-length TGF-alpha protein (Uniprot: P01135) | |
| 0.1 mL | |
| Angiogenesis, Cell Cycle and Replication, Signal Transduction | |
| 7039 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| TGFA/1119 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction