missing translation for 'onlineSavingsMsg'
Learn More

TG4-155, MedChemExpress

Product Code. 16457566
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457566 100 mg 100mg
16443626 25 mg 25mg
16437668 10 mg 10mg
16433677 50 mg 50mg
16400138 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16457566 Supplier Medchem Express Supplier No. HY18971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TG4-155 is a potent, brain-permeant and selective EP2 receptor antagonist with a Ki of 9.9 nM. TG4-155 shows low nanomolar antagonist activity against only EP2 and DP1. TG4-155 has an EP2 Schild KB of 2.4 nM and displays 550-4750-fold selectivity for EP2 over EP1, EP3, EP4 and IP, but only 14-fold selectivity against the DP1 receptor.

TG4-155 is a potent, brain-permeant and selective EP2 receptor antagonist with a Ki of 9.9 nM. TG4-155 shows low nanomolar antagonist activity against only EP2 and DP1. TG4-155 has an EP2 Schild KB of 2.4 nM and displays 550-4750-fold selectivity for EP2 over EP1, EP3, EP4 and IP, but only 14-fold selectivity against the DP1 receptor.

Specifications

Chemical Name or Material TG4-155
CAS 1164462-05-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H26N2O4
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (316.89 mM; Need ultrasonic)
SMILES O=C(NCCN1C(C)=CC2=C1C=CC=C2)/C=C/C3=CC(OC)=C(OC)C(OC)=C3
Molecular Weight (g/mol) 394.46
Formula Weight 394.46
Percent Purity 99.69%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.