missing translation for 'onlineSavingsMsg'
Learn More

TG101209, MedChemExpress

Product Code. 16453958
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453958 10 mg 10mg
16436677 5 mg 5mg
16434976 100 mg 100mg
16410317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16453958 Supplier Medchem Express Supplier No. HY10410
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TG101209 is a selective JAK2 inhibitor with IC50 of 6 nM, less potent to Flt3 and RET with IC50 of 25 nM and 17 nM, appr 30-fold selective for JAK2 than JAK3, and sensitive to JAK2V617F and MPLW515L/K mutations.

TG101209 is a selective JAK2 inhibitor with IC50 of 6 nM, less potent to Flt3 and RET with IC50 of 25 nM and 17 nM, appr 30-fold selective for JAK2 than JAK3, and sensitive to JAK2V617F and MPLW515L/K mutations.

Specifications

Chemical Name or Material TG101209
CAS 936091-14-4
Color Gray
Purity Grade Notes Research
Molecular Formula C26H35N7O2S
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (98.10 mM)
SMILES O=S(C1=CC=CC(NC2=NC(NC3=CC=C(C=C3)N4CCN(CC4)C)=NC=C2C)=C1)(NC(C)(C)C)=O
Molecular Weight (g/mol) 509.67
Formula Weight 509.67
Percent Purity 98.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.