missing translation for 'onlineSavingsMsg'
Learn More

TG 100572 Hydrochloride, MedChemExpress

Product Code. 16420007
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16420007

Brand: Medchem Express HY10185

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TG 100572 Hydrochloride is a multi-targeted kinase inhibitor which inhibits receptor tyrosine kinases and Src kinases; has IC50s of 2, 7, 2, 16, 13, 5, 0.5, 6, 0.1, 0.4, 1, 0.2 nM for VEGFR1, VEGFR2, FGFR1, FGFR2, PDGFRβ, Fgr, Fyn, Hck, Lck, Lyn, Src, Yes, respectively.

TG 100572 Hydrochloride is a multi-targeted kinase inhibitor which inhibits receptor tyrosine kinases and Src kinases

Specifications

Chemical Name or Material TG 100572 Hydrochloride
CAS 867331-64-4
Color Red
Purity Grade Notes Research
Molecular Formula C26H27Cl2N5O2
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (48.79 mM; Need ultrasonic)
SMILES ClC1=CC=C(O)C=C1C2=CC(C)=C(N=C(NC(C=C3)=CC=C3OCCN4CCCC4)N=N5)C5=C2.Cl
Molecular Weight (g/mol) 512.43
Formula Weight 512.43
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.