missing translation for 'onlineSavingsMsg'
Learn More

Ebro Electronic™ TFX 430 Precision Thermometer
GREENER_CHOICE

Product Code. 10399282
Click to view available options
Probe Type:
Blunt
Glass Coated
Pointed
Without probe
Length (Metric):
190 mm
355 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10399282

Brand: Ebro Electronic™ 13405430A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reference thermometer with exchangeble PT 100 probe

TFX 430 Model Precision Thermometer. Powered by a replaceable lithium battery, these precision thermometers are handy, watertight temperature measurement devices for a whole range of measurement and control applications in the laboratory and in industry.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; EN 13485
Disposable Single-use
For Use With (Application) Applications: Laboratory, Temperature monitoring, Measurement of air, fluids, plastic materials, aggressive mediums and surfaces, Reference thermometer
Height (Metric) 38 mm
Includes Thermometer, calibration certificate, instruction manual
Length (Metric) 190 mm
Material Plastic
Model TFX 430
Probe Type Without probe
Resolution 0.01°C from −10.00°C to 199.99°C, 0.1°C remaining range
Weight (Metric) 0.24 kg
Product Type Precision Thermometer
Data Storage None
Max. Temperature (English) 932°F
Max. Temperature (Metric) 500°C
Min. Temperature (English) -148°F
Min. Temperature (Metric) -100°C
Temperature Range (Metric) -100°C to +500°C
Calibrated Yes
For Use With (Equipment) AG 130, AG 140, AG 150, AX 110, TPX 130, TPX 230, TPX 330
Battery Life Approx. 5 Years
Battery Type Lithium 3 V/Typ CR 2477
Accuracy ±0.05°C at -50° to +199.99°C; ±0.2°C for the remaining measuring range
Display LCD
Width (Metric) 120 mm
IP Rating IP67
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.