missing translation for 'onlineSavingsMsg'
Learn More

Ebro Electronic™ TFX Core Thermometer Series
GREENER_CHOICE

Product Code. 10535902
Click to view available options
Probe Type:
Blunt
Fixed
Glass Coated
Pointed
Pointed, Pt 1000 sensor
Includes:
Calibration Cert., No Cable
No Cable
PTFE Cable, Calibration Cert.
Silicone Cable
Silicone Cable, Calibration Cert.
Model:
TFX 410
TFX 410-1 & TPX 200
TFX 410-1 & TPX 400-150
TFX 410-1 TPX 400
TFX 410-1 and TPX 100
TFX 410-1 and TPX 200-40
TFX 410-1 and TPX 300
TFX 410-1 and TPX 440
TFX 420 + TPX 200
TFX 420 + TPX 400
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10535902

Brand: Ebro Electronic™ 13405419A

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Precise and very robust thermometer with a big display

Thermometer for various applications in food, laboratory, industial use. All thermometers includes a factory calibration certificate. The thermometer measures with PT 1000 temperature sensor.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; EN 13485; (EC) 1935/2004 Materials and articles which come into contact with foodstuffs; The Food and Feed Hygiene and Safety Regulations 2020 2020 No. 1504; PART 3; Regulation 16
Disposable Single-use
For Use With (Application) For bakeries, butcheries and for the food industry
Height (Metric) 54 mm
Includes Calibration Cert., No Cable
Length (Metric) 355 mm
Material Stainless Steel, Plastic
Model TFX 410-1 and TPX 300
Probe Type Glass Coated
Resolution 0.1 °C
Weight (Metric) 0.4 kg
Product Type Precision Core Thermometer
Data Storage None
Description Without cable
Max. Temperature (English) 572°F
Max. Temperature (Metric) 300°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (Metric) -50°C to +300°C
Calibrated Yes
For Use With (Equipment) AG 130, AG 140, AG 150, AG 160, AX 100
Battery Life Approx. 5 Years
Battery Type Lithium 3 V/Typ CR 2477
Accuracy ±0.3°C/±0.5°F
Immersion Length 190 mm
Display LCD
Width (Metric) 162 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.