missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TfR (Transferrin R) Antibody (TFRC/1149), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11596F
This item is not returnable.
View return policy
Description
TfR (Transferrin R) Monoclonal antibody specifically detects TfR (Transferrin R) in Human samples. It is validated for Western Blot
Specifications
| TfR (Transferrin R) | |
| Monoclonal | |
| FITC | |
| CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
| Recombinant human TfR (Transferrin R) protein (Uniprot: P02786) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 7037 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot | |
| TFRC/1149 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction