missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TfR (Transferrin R) Antibody (MEM-189), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
TfR (Transferrin R) Monoclonal antibody specifically detects TfR (Transferrin R) in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | TfR (Transferrin R) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MEM-189 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr |
| Host Species | Mouse |
| Immunogen | KG1 human acute myelogenous leukaemia cell line (NM_001128148.1). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?