missing translation for 'onlineSavingsMsg'
Learn More

TfR (Transferrin R) Antibody (30002) [Biotin], Novus Biologicals™

Product Code. 30737162 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737162 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737162 Supplier Novus Biologicals Supplier No. FAB24741B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TfR (Transferrin R) Monoclonal antibody specifically detects TfR (Transferrin R) in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TfR (Transferrin R)
Applications ELISA
Classification Monoclonal
Clone 30002
Conjugate Biotin
Formulation PBS
Gene Alias CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human TfR (Transferrin R) protein, Cys89-Phe760, Accession # CAA25527
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7037
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.