missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | TFPT |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18363064
|
Novus Biologicals
NBP3-04589-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18395245
|
Novus Biologicals
NBP3-04589-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFPT Polyclonal specifically detects TFPT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| TFPT | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 29844 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| amida, INO80 complex subunit F, partner of the E2A, Protein FB1, TCF3 (E2A) fusion partner (in childhood Leukemia), TCF3 fusion partner | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-253 of human TFPT (NP_037474.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title