missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TFPI Antibody, R&D Systems™
Rat Monoclonal Antibody
£332.60
Specifications
Antigen | TFPI |
---|---|
Clone | 372311 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
TFPI Monoclonal specifically detects TFPI in Mouse samples. It is validated for Western Blot.Specifications
TFPI | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
O54819 | |
7035 | |
Mouse myeloma cell line NS0-derived recombinant mouse TFPI Leu29-Lys289 Accession # O54819 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse TFPI in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows no cross-reactivity with recombinant human (rh) TFPI or rmTFPI-2. |
372311 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
convertin, EPI, EPITFPI1LACITFI, Extrinsic pathway inhibitor, LACI, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
TFPI | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only