missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPI Rabbit anti-Human, Clone: 3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89568-100UL
This item is not returnable.
View return policy
Description
TFPI Monoclonal specifically detects TFPI in Human samples. It is validated for ELISA.
Specifications
| TFPI | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 7035 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 3 | |
| ELISA 1:1000-1:2000 | |
| convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
| Recombinant Human TFPI Protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction