missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFPI Monoclonal Antibody (OTI4H9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526775
This item is not returnable.
View return policy
Description
TFPI Monoclonal Antibody for Western Blot, IHC (P)
Tissue factor pathway inhibitor (TFPI), also known as lipoprotein-associated coagulation inhibitor, is a protease inhibitor that regulates the tissue factor (TF)-dependent pathway of blood coagulation. TFPI is glycosylated and predominantly found in the vascular endothelium and plasma in both free forms and complexed with plasma lipoproteins. The coagulation process initiates with the formation of a factor VIIa-TF complex, which proteolytically activates additional proteases (factors IX and X) and ultimately leads to the formation of a fibrin clot. TFPI inhibits the activated factor X and VIIa-TF proteases in an autoregulatory loop.
Specifications
| TFPI | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P10646 | |
| TFPI | |
| Human recombinant protein fragment corresponding to amino acids 29-304 of TFPI produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H9 | |
| Unconjugated | |
| TFPI | |
| A630013F22Rik; anti-convertin; AW552122; EPI; Extrinsic pathway inhibitor; LACI; Lipoprotein-associated coagulation inhibitor; OTTHUMP00000163472; OTTHUMP00000205431; TFI; TFPI; TFPI1; Tissue factor pathway inhibitor; tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7035 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction