missing translation for 'onlineSavingsMsg'
Learn More

TFPI Antibody (401), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18344621 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344621 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344621 Supplier Novus Biologicals Supplier No. NBP289570AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TFPI Monoclonal specifically detects TFPI in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFPI
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA
Classification Monoclonal
Clone 401
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TFPI (NP_006278.1; Met1-Lys282).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7035
Test Specificity Has cross-reactivity in ELISA with: Mouse TFPI No cross-reactivity in ELISA with: Human TFPI2 Human Vitronectin
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.