missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TFPI Monoclonal antibody specifically detects TFPI in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | TFPI |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3 |
| Conjugate | DyLight 594 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human TFPI (NP_006278.1; Met 1-Lys 282). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?