missing translation for 'onlineSavingsMsg'
Learn More

TFPI-2 Antibody (OTI1D4), DyLight 594, Novus Biologicals™

Product Code. 18413576 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413576 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413576 Supplier Novus Biologicals Supplier No. NBP274494DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TFPI-2 Monoclonal antibody specifically detects TFPI-2 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFPI-2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1D4
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Placental protein 5, PP5FLJ21164, TFPI-2REF1, tissue factor pathway inhibitor 2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 23-235 of human TFPI-2 (NP_006519) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7980
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.