missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPI-2 Antibody (MM0564-9F6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£171.00 - £444.00
Specifications
| Antigen | TFPI-2 |
|---|---|
| Clone | MM0564-9F6 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18017126
|
Novus Biologicals
NBP2-11918-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18059816
|
Novus Biologicals
NBP2-11918 |
0.1 mg |
£444.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFPI-2 Monoclonal specifically detects TFPI-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| TFPI-2 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| P48307 | |
| 7980 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0564-9F6 | |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Placental protein 5, PP5FLJ21164, TFPI-2REF1, tissue factor pathway inhibitor 2 | |
| TFPI2 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title