missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPI-2 Antibody (MM0564-9F6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11918AF350
This item is not returnable.
View return policy
Description
TFPI-2 Monoclonal specifically detects TFPI-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| TFPI-2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TFPI2 | |
| Human recombinant TFPI-2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MM0564-9F6 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Placental protein 5, PP5FLJ21164, TFPI-2REF1, tissue factor pathway inhibitor 2 | |
| Mouse | |
| Protein G purified | |
| Cardiovascular Biology | |
| 7980.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction