missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress TFLLR-NH2(TFA)

Product Code. 16402699
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402699 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402699 Supplier MedChemExpress Supplier No. HYP0226A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress TFLLR-NH2 (TFA) is a selective PAR1 agonist with an EC50 of 1.9 μM.

Specifications

Purity 99.77%
CAS 1313730-19-6
Content And Storage Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 761.83 g/mol
Product Type Peptides
Quantity 1 mg
Shipping Condition Room temperature
Solubility DMSO : 100 mg/mL (131.26 mM∣Need ultrasonic) ∣H2O : 100 mg/mL (131.26 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.