missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14436-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein component of the spliceosome that promotes the release of the lariat-intron during late-stage splicing through the recruitment of a pre-mRNA splicing factor called DEAH-box helicase 15. The encoded protein contains a G-patch domain, a hallmark of RNA-processing proteins, that binds DEAH-box helicase 15. This protein contains an atypical nuclear localization sequence as well as a nuclear speckle-targeting sequence, enabling it to localize to distinct speckled regions within the cell nucleus. Polymorphisms in this gene are associated with dental caries suggesting a role in amelogenesis. Alternative splicing results in multiple transcript variants.Specifications
| TFIP11 | |
| Polyclonal | |
| Unconjugated | |
| TFIP11 | |
| bK445C9.6, STIP, STIP 1, TFIP11, TIP39 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24144, 288718, 54723 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5U2Y6, Q9ERA6, Q9UBB9 | |
| Tfip11 | |
| TFIP11 Fusion Protein Ag5821 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction